Adapting well known chromatographic techniques for the analysis of biomolecules
Webinar
Thursday, 18th June, 2020  
15:00 to 16:00 CEST (Berlin, Paris, Madrid)
Register
Regulatory guidelines (ICH Q6B) state that a detailed characterisation of all therapeutic biomolecules is carried out prior to any new product launch. It is important to confirm primary structure and identify any post translational modifications (PTMs) which have occurred during manufacturing processes. When considering biosimilars or biobetters, a comparison should be made with the Originator therapeutic.
Chromatography is a well established technique for the analysis of small molecule therapeutics, their related substances and stability, and is useful for both quantitation and quality purposes. In this webinar we discuss how you can draw from experience with small molecule method development to optimise biomolecule methodologies. We also discuss how ion-pair reagents can be added to the mobile phase to enhance the interaction of more polar biomolecules like oligonucleotides, and lead to excellent separations even to the n+/-1 level. Whether you are working with peptides or proteins, this webinar aims to provide ideas on how to approach your analysis using well understood and recognised modes of chromatography.