Understanding analyte physicochemical properties and their value in LC method development
Webinar
Thursday, May 12, 2022
3:00 PM - 4:00 PM EDT (USA, Canada)
Replay
Analyte retention in liquid chromatography (LC) is driven by molecular interactions between analyte molecules and both the mobile phase and stationary phase. The type and extent of these interactions are dependent on the physicochemical properties of the analyte. An understanding of analyte properties is invaluable for method development, enabling selection of the appropriate chromatographic mode and key method parameters (e.g. mobile phase and stationary phase selection). For example, analyte pKa, logP and logD values can be used to evaluate ionisation state and hydrophobicity and can help predict analyte retention and guide mobile phase composition, pH and buffer selection. This webinar will review the range of analyte properties that are important to consider when developing methods. These will be discussed in the context of the interactions between the analyte, mobile phase and stationary phase, which defines the separation process. Additionally, analyte structure presents opportunities to optimise separation selectivity by exploiting additional retention mechanisms offered by novel stationary phase chemistries. This will be discussed with examples demonstrating the impact this can have on the chromatographic separation obtained.
For more information,please contact us at webinar@avantorsciences.com