Puritan Medical Products
Puritan Medical Products Co., LLC, is over 100 years old and is a family-owned American company known worldwide as the trusted manufacturer of single-use products for controlled environments, diagnostics, specimen collection, environmental, forensics, genetics, medical, and microbiology media industries. Puritan is FDA-registered and ISO-certified. Puritan's products help customers collect, analyze, and transport specimens with the utmost confidence in the products’ integrity. All patented flock swabs and related products are manufactured in its Guilford, Maine and Pittsfield, Maine facilities.
Puritan's patented HydraFlock® and PurFlock Ultra® flocked swabs have optimum absorption and elution capabilities and are the preferred swab for specimen collection and patient comfort. UniTranz-RT® (Universal transport media) is used to test for viruses, chlamydia, mycoplasma, and ureaplasma specimens at room temperature (RT) or long-term freezer storage. Puritan’s Opti-Swab® Modified Liquid Amies transport medium maintains viability of aerobic, anaerobic and fastidious bacteria.
Puritan's media transport systems combined with high-performing swabs create the ideal product for specimen collection and transport. Puritan will always be in the forefront, meeting the needs of emerging technologies.
HydraFlock® and PurFlock Ultra® Specimen Collection Devices
HydraFlock provides superior collection and a quick release of specimens, enhanced by its unique microstructure with multi-length fibers. PurFlock Ultra’s high purity features make it an ideal swab for rapid diagnostic tests.
COVID-19 Testing Supplies
When conducting diagnostic testing for COVID-19, CDC recommends Nasopharyngeal (NP) or Oropharyngeal (OP) swabbing. Puritan's foam or flock swabs are the ideal swab for nasopharyngeal and oropharyngeal COVID-19 specimen collection.
Media Transport Systems
Puritan's media transport systems combined with high-performing swabs create the ideal product for specimen collection and transport.
Cleanroom and Industrial Swabs
In controlled environments, precision is everything. You can count on Puritan for cleanroom and industrial swabs that are manufactured in an ISO:2008 certified and CE-compliant facility, ensuring the quality of your end product.