Hypersil GOLD™ Columns by Thermo Scientific
Thermo Scientific™ Hypersil GOLD™ columns offers chromatographers outstanding peak shape for reversed phase, ion exchange, HILIC or normal phase chromatography.
Hypersil Gold HPL and UHPLC Columns.
Outstanding peak shape for your separations. Hypersil Gold columns are an ideal choice for new method development, delivering outstanding peak shape and increased sensitivity for your separations.
Endcapped, ultra-pure, silica-based columns with exceptional peak shape and resolution for HPLC and LC-MS