Human Recombinant HBEGF
Supplier: STEMCELL Technologies
78163 78163.1
MSPP-78163EA 130.08 USD
MSPP-78163 MSPP-781631006
Human Recombinant HBEGF
Proteins and Peptides
Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a member of the EGF family (Nishi and Klagsbrun). HBEGF promotes blastocyst adhesion to the uterine wall (Iwamoto and Mekada). It also plays a role in smooth muscle cell hyperplasia and brain injury (Nishi and Klagsburn). HBEGF produced by CD4+ T cells promotes wound healing by stimulating migration and proliferation of keratinocytes, fibroblasts, and smooth muscle cells (Blotnick et al.). It binds to EGFR, ErbB4, ErbB2, and ErbB3, activating the PI3K/AKT signaling cascade (Iwamoto and Mekada). HBEGF is produced in a variety of cells, where it contributes to physiological and pathological processes. HBEGF is overexpressed in ovarian, breast, gastric, colorectal, pancreatic, and endometrial cancers, which likely contributes to pathogenesis (Miyata et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').