Human Recombinant Myostatin
Supplier: STEMCELL Technologies
78119 78119.1
MSPP-78119EA 304 USD
MSPP-78119 MSPP-781191006
Human Recombinant Myostatin
Proteins and Peptides
Myostatin is a member of transforming growth factor beta (TGF-β) superfamily that signals by binding to activin type II receptor, resulting in the recruitment of either ALK3 or ALK4 coreceptor, and activation of SMAD2/3 (Kondás et al.). It is released by myocytes and inhibits muscle growth and ability to regenerate (Lee and Lee). Myostatin appears to affect the lipid catabolic metabolism of adipocytes and inhibits preadipocyte differentiation, as demonstrated in the 3T3-L1 cell line (Li et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').