Mouse Recombinant IL-5
Supplier: STEMCELL Technologies
78049 78049.1
MSPP-78049EA 955.85 USD
MSPP-78049 MSPP-780491006
Mouse Recombinant IL-5
Proteins and Peptides
Interleukin 5 (IL-5) is a member of the short-chain 4-α-helical bundle subset of hematopoietic cytokines. It binds to the specific α receptor, which in turn recruits the common β receptor that initiates signaling via JAK/STAT and MAPK pathways. IL-5 is produced by Th2 cells, eosinophils, activated mast cells, and ILC2 cells. IL-5 affects the differentiation, activation, and survival of eosinophils, thus playing an important role in allergic inflammation, asthma, and parasite immunity. Stimulation of eosinophils with IL-5 leads to their activation, upregulation of CD11b expression, and inhibition of apoptosis (Shearer).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').