Human Recombinant Follistatin
Supplier: STEMCELL Technologies
78109.1 78109
MSPP-781091006EA 1240.85 USD
MSPP-781091006 MSPP-78109
Human Recombinant Follistatin
Proteins and Peptides
Follistatin, a glycosylated monomeric protein, is a modulator of transforming growth factor beta (TGF-β) superfamily signaling. It binds to and inhibits the function of activin, myostatin, growth differentiation factors, and bone morphogenetic proteins (BMP) (Hansen and Plomgaard). Follistatin inhibits mesoderm induction, suppresses synthesis and secretion of pituitary follicle-stimulating hormone, regulates liver regeneration, and causes infertility (Guo et al.; Iemura et al.). Follistatin exhibits anti-inflammatory effects, and it could be used as a biomarker in cancer (Hansen and Plomgaard).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').