Mouse Recombinant IL-7
Supplier: STEMCELL Technologies
78054 78054.1
MSPP-78054EA 1075.69 USD
MSPP-78054 MSPP-780541006
Mouse Recombinant IL-7
Proteins and Peptides
Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette and Surh), and provides signals to promote survival, proliferation, and differentiation during B cell development (Clark et al.; Corfe and Paige).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').