Rat IGF-I
Supplier: Rockland Immunochemical
012-001-V21-0010 012-001-V21-0050
75928-950EA 2062 USD
75928-950 75928-954 75934-658
Rat IGF-I
Proteins and Peptides
Recombinant Rat IGF-I control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Insulin-like Growth Factor I, IGF-I, is a growth factor produced in response to growth hormone-stimulated liver tissue and can be found circulating throughout the body and throughout life. IGF-I activates the IGF-I receptor (IGF1R) and the insulin receptor to mediate growth of almost every cell of the body. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Mature human IGF-I is 100% homologous with bovine and porcine proteins. Recombinant rat IGF-I is a non-glycosylated protein, containing 70 amino acids, with a molecular weight of 7.7 kDa.
Insulin-like Growth Factor I Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Insulin-like Growth Factor I in immunological assays.