Mouse Recombinant VEGF-120
Supplier: Rockland Immunochemical
Recombinant Mouse VEGF-120 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several variants, one being VEGF-120. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%. Recombinant mouse VEGF-120 is an alternative spliced version of mouse VEGF-165. It is a non-glycosylated homodimer, containing two chains, with a molecular weight of 28.2 kDa.
Vascular Endothelial Growth Factor-120 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Vascular Endothelial Growth Factor-120 in immunological assays.