Mouse SCF
Supplier: Rockland Immunochemical
010-001-W20-0010 010-001-W20-0100
75932-254EA 4582 USD
75932-254 75932-260 75932-266 75932-272
Mouse SCF
Proteins and Peptides
Recombinant Mouse SCF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Stem Cell Factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor known as c-Kit (CD117) and is thought to play a critical role in the maintenance or survival of hematopoietic stem cells. Human SCF shows no activity on murine cells, but murine and rat SCF are active on human cells. Recombinant mouse SCF is a non-glycosylated protein, containing 164 amino acids, with a molecular weight of 18.3 kDa.
Stem Cell Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Stem Cell Factor in immunological assays.