Mouse Exodus-2 / CCL21
Supplier: Rockland Immunochemical
010-001-U75-0100 010-001-U75-0020 010-001-U75-0005
10802-646EA 4582 USD
10802-646 10802-648 10802-650 10802-652
Mouse Exodus-2 / CCL21
Proteins and Peptides
Recombinant Mouse Exodus-2 / CCL21 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Exodus-2, also known as CCL21 and 6Ckine, is a novel chemokine produced at high levels in human lymph node tissue and the spleen. 6Ckine signals through the CCR7 receptor to recruit thymocytes and activated T cells in vitro, but is not known to be chemotactic for monocytes. Human and mouse Exodus-2 share > 85% amino acid identity. Recombinant mouse Exodus-2 is a non-glycosylated protein, containing 110 amino acids, with a molecular weight of 12 kDa.
Exodus-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Exodus-2 in immunological assays.