Human Recombinant TGF-Beta1
Supplier: Rockland Immunochemical
009-001-W26-0100 009-001-W26-0005
75933-058EA 1920.3 USD
75933-058 75933-060
Human Recombinant TGF-Beta1
Proteins and Peptides
Recombinant Human TGF-Beta1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Transforming Growth Factor-beta 1 (TGF-β1) is a member of the TGF-beta superfamily. Members of this family are known to exhibit regulatory activity in immunity and proliferation pathways. TGF-β1 signals through SMAD proteins via the TGF-β1 RI and RII receptors. Mature human TGF-β1 is a glycosylated homodimer, containing two 112 amino acids chains, with a total molecular weight of 25 kDa.
Transforming Growth Factor beta 1 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Transforming Growth Factor beta 1 in immunological assays.