Human Recombinant MIF
Supplier: Rockland Immunochemical
Recombinant Human MIF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Migration Inhibitory Factor (MIF) is a pro-inflammatory cytokine that acts on fibroblasts by inducing IL-1, IL-8 and MMP expression. MIF stimulates NO production and TNF-α release following IFNγ-activation of macrophages. Recombinant human MIF is a non-glycosylated protein, containing 115 amino acids, with a molecular weight of 12.5 kDa.
Migration Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Migration Inhibitory Factor in immunological assays.