Human Recombinant IL-16
Supplier: Rockland Immunochemical
Recombinant Human IL-16 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Interleukin 16 (IL-16) is produced primarily by CD4+ and CD8+ T cells and acts as a chemo-attractant for lymphocytes, monocytes, eosinophils, dendritic cells and Langerhans cells. Additionally, IL-16 has been reported to upregulate IL-2 receptor (CD25), induce progression of cells to the G1 phase and suppress HIV & SIV replication. Recombinant human IL-16 is a non-glycosylated protein, containing130 amino acids, with a molecular weight of 13.5 kDa.
Interleukin-16 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-16 in immunological assays.