p73 alpha protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S70 009-001-S70S
75934-044EA 714 USD
75934-044 75934-042
p73 alpha protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p73 alpha control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p73α is a member of the p53 family of transcription factors that are involved in cellular responses to stress and development. P73 has high sequence similarity to p53 and p63, which allows p63 and p73 to transactivate p53-responsive genes causing cell cycle arrest and apoptosis. P73 is a stress-response gene and a downstream effector in the p53 pathway (1). The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. p73 is the components of a mismatch repair-dependent apoptosis pathway which contributes to cisplatin-induced cytotoxicity (2). p73α Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
p73α Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p73α Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~110kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.