Mouse Recombinant Il7 (from E. coli)
Supplier: Rockland Immunochemical
010-001-B94-0020 010-001-B94-0100
75929-176EA 352.09 USD
75929-176 75929-178
Mouse Recombinant Il7 (from E. coli)
Proteins and Peptides
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cell s, and natural killer cells. Human and mouse IL-7 show species cross-reactivity. Recombinant mouse IL-7 is a non-glycosylated protein, containing 172 amino acids, with a molecular weight of 17.4 kDa.