Human Recombinant CSF3 (from E. coli)
Supplier: Rockland Immunochemical
009-001-U93-0010 009-001-U93-0100
10801-550EA 6248 USD
10801-550 10801-552 10801-554 10801-556
Human Recombinant CSF3 (from E. coli)
Proteins and Peptides
Granulocyte-Colony Stimulating Factor (G-CSF) is a growth factor that is considered the most potent inducer of terminal differentiation to granulocytes and macrophages of leukemic myeloid cell lines. The synthesis of G-CSF can be induced by bacterial endotoxins, TNF, IL-1 and GM-CSF. Prostaglandin E2 inhibits the synthesis of G-CSF, while in epithelial, endothelial, and fibroblastic cells, secretion of G-CSF is induced by IL-17. Human and mouse G-CSF are cross-reactive. Recombinant human G-CSF is a non-glycosylated protein, containing 175 amino acids, with a molecular weight of 18.8 kDa.