[Ala8]-Humanin, [Ala8]-HN, sHNA, Anaspec Inc.

Supplier: Anaspec Inc
AS-60888
103006-056EA 394.62 USD
103006-056
[Ala8]-Humanin, [Ala8]-HN, sHNA, Anaspec Inc.
Proteins and Peptides
Protection activity of humanin (HN) against neuronal cell death is abrogated in this peptide, where Cys8 is substituted by Ala.
Sequence:MAPRGFSALLLLTSEIDLPVKRRA
MW:2655.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR