Matrix Metalloproteinase-19 (MMP-19) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK307-0001
76002-364EA 756 USD
76002-364
Matrix Metalloproteinase-19 (MMP-19) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Assays Protein Assays
Designed to screen MMP-19 inhibitors using a quenched fluorogenic peptide.
The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-19, a potential therapeutic target. The compound NNGH is also included as a prototypic control inhibitor.
Ordering information: Contains 1 vial MMP-19 enzyme, 1 vial Substrate (OMNIMMP® RED), 1 vial 6’-TAMRA calibration standard, 1 vial control inhibitor (NNGH), 1 bottle (20 mL) assay buffer, 1 black 96-well microplate, Instructions.