Mouse Recombinant CSF3 (from CHO cells)
Supplier: ADIPOGEN CORP MS
CHI-MF-110CSF3-C010 CHI-MF-110CSF3-C050
102981-540EA 249.23 USD
102981-540 102981-542
Mouse Recombinant CSF3 (from CHO cells)
Proteins and Peptides
G-CSF is a pleiotropic cytokine best known for its specific effects on the proliferation, differentiation and activation of hematopoietic cells of the neutrophilic granulocyte lineage. It is produced mainly by monocytes and macrophages upon activation by endotoxin, TNF-alpha and IFN-gamma. Other cell types including fibroblasts, endothelial cells, astrocytes and bone marrow stromal cells can also secrete G-CSF after LPS, IL-1 or TNF-alpha activation. In addition, various carcinoma cell lines and myeloblastic leukemia cells can express G-CSF constitutively. G-CSF stimulates growth, differentiation and functions of cells from the neutrophil lineage in vitro. Consistent with its in vitro functions, G-CSF has been found to play important roles in defense against infection, in inflammation and repair and in the maintenance of steady state hematopoiesis. Recombinant human G-CSF has been approved for the amelioration of chemotherapy induced neutropenia as well as for severe chronic neutropenia following marrow transplant.