Human Recombinant CD278 (from CHO Cells)
Supplier: ADIPOGEN CORP MS
CHI-HF-210ICOS-C025 CHI-HF-210ICOS-C100
102981-524EA 228.46 USD
102981-524 102981-526
Human Recombinant CD278 (from CHO Cells)
Proteins and Peptides
CD278, also called inducible costimulator (ICOS) or CRP1 (CD28-related protein-1), is a member of the growing CD28 family of immune costimulatory receptors. Other family members are CD28, CTLA-4 and PD-1. ICOS is expressed on most CD45RO+ cells. ICOS expression is upregulated within approximately 24-48 hours of activation on Th primed cells. B7-H2, a member of the B7 family of costimulatory ligands, has been identified as the ICOS ligand. The B7-H2/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.