Human Recombinant Myeloperoxidase
Supplier: Enzo Life Sciences
200-600A-U025
200-600A-5025
76001-902EA
288.85
USD
76001-902
76001-904
Human Recombinant Myeloperoxidase
Proteins and Peptides
Isolated from human leukocytes. Negative for HBsAG-, HCV- and HIV-antibodies.££Haem protein that catalyzes oxidations by H2O2, including MPO-chloride-mediated killing of microbes and tumor cells, inactivation of chemotactic factors, cross-linking of proteins and iodination of proteins.