Z-Leu-Leu-Leu-Amc
Supplier: ADIPOGEN CORP MS
AG-CP3-0019-M001 AG-CP3-0019-M005
102988-488EA 55.38 USD
102988-488 102988-490
Z-Leu-Leu-Leu-Amc
Proteins and Peptides
Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome. Excitation: 345nm. Emission: 445nm. This substrate is useful for inhibitor screening and kinetic analysis.
  • Soluble in DMSO.