Human Recombinant Resistin (from E. coli)
Supplier: Peprotech
450-19-5UG 450-19-25UG 450-19-50UG 450-19-100UG 450-19-250UG 450-19-500UG 450-19-1MG
10781-332EA 132.81 USD
10781-332 10781-334 10781-336 10781-338 10781-340 10781-342 10781-344
Human Recombinant Resistin (from E. coli)
Proteins and Peptides
Resistin belongs to a family of tissue-specific cytokines termed FIZZ (found in inflammatory zones) and RELM. The four known members of this family, resistin, RELMα, RELMβ, and RELMγ, share a highly conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. Resistin is an adipose-derived cytokine (adipokine) whose physiological function and molecular targets are largely unknown. Studies have shown that resistin suppresses insulin's ability to stimulate glucose uptake, and postulated that resistin might be an important link between obesity and Type 2 diabetes. Other studies have indicated that resistin expression is severely suppressed in obesity, and that it may act as a feedback regulator of Adipogenesis. Recombinant Human Resistin is a 19.5 kDa, disulfide-linked, homodimeric protein composed of two identical 92 amino acid chains linked by a single disulfide bond.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.