Rat Recombinant IL-5 (from E. coli)
Supplier: Peprotech
400-05-2UG 400-05-10UG 400-05-50UG 400-05-100UG 400-05-500UG 400-05-1MG
10780-910EA 132.81 USD
10780-910 10780-912 10780-914 10780-916 10780-920 10780-922
Rat Recombinant IL-5 (from E. coli)
Proteins and Peptides
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 is cross-species reactive. Recombinant Rat IL-5 is a 26.0 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.