Human Recombinant IL-3 (from E. coli)
Supplier: Peprotech
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoiesis, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is a species-specific, variably glycosylated cytokine. Recombinant Human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.