Human Recombinant SPARC/Osteonectin (from CHO Cells)
Supplier: Peprotech
120-36-500UG 120-36-1MG
10779-184EA 1445.88 USD
10779-184 10779-186
Human Recombinant SPARC/Osteonectin (from CHO Cells)
Proteins and Peptides
SPARC/Osteonectin is a secreted, evolutionarily-conserved, collagen-binding glycoprotein that is involved in a variety of cellular activities. It is highly expressed in tissues undergoing morphogenesis, remodeling and wound repair. SPARC/Osteonectin and its related peptides bind to numerous proteins of the extracellular matrix (ECM), affect ECM protein expression, influence cellular adhesion and migration, and modulate growth factor-induced cell proliferation and angiogenesis. SPARC/Osteonectin consists of three domains: an N-terminal acidic region that binds calcium ions with low affinity, a module containing two EF-hand motifs that bind calcium with high affinity, and a cysteine-rich follistatin-like domain. Recombinant Human SPARC/Osteonectin is a glycoprotein containing 286 amino acids that migrates at an apparent MW of 43.7 kDa by SDS-PAGE analysis due to the effect of glycosylation. The calculated molecular weight of Recombinant Human SPARC/Osteonectin is 32.7 kDa.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.