Mouse Recombinant MIP-3alpha (from E. coli)
Supplier: Peprotech
250-27-5UG 250-27-20UG 250-27-100UG 250-27-250UG 250-27-500UG 250-27-1MG
10773-358EA 139.89 USD
10773-358 10773-360 10773-362 10773-364 10773-366 10773-368
Mouse Recombinant MIP-3alpha (from E. coli)
Proteins and Peptides
MIP-3α is a CC chemokine, expressed in the liver, lymph nodes, appendix, PBL and lungs that can signal through the CCR6 receptor. MIP-3α is chemotactic towards lymphocytes and dendritic cells. Additionally, it promotes the adhesion of memory CD4+ T cells, and inhibits colony formation of bone marrow myeloid immature progenitors. Recombinant Murine MIP-3α (CCL20) is a 7.9 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.