Mouse Recombinant Eotaxin (from E. coli)
Supplier: Peprotech
250-01-250UG 250-01-500UG 250-01-1MG
10773-114EA 1615.45 USD
10773-114 10773-116 10773-118
Mouse Recombinant Eotaxin (from E. coli)
Proteins and Peptides
Eotaxin is a CC chemokine that signals through the CCR3 receptor. It is produced by IFN-γ-stimulated endothelial cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and, along with Eotaxin-2 and Eotaxin-3, plays a key role in the regulation of eosinophil recruitment in the asthmatic lung and in allergic reactions. Recombinant Murine Eotaxin (CCL11) is an 8.4 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.