Human Recombinant IL-11 (from E. coli)
Supplier: Peprotech
200-11-2UG 200-11-10UG 200-11-100UG 200-11-250UG 200-11-500UG 200-11-1MG
10772-998EA 139.89 USD
10772-998 10773-000 10773-002 10773-004 10773-006 10773-008
Human Recombinant IL-11 (from E. coli)
Proteins and Peptides
IL-11 is a multifunctional cytokine produced by stromal cells, such as fibroblasts, epithelial cells and osteoclasts. It is expressed in a wide variety of tissues, including thymus, lung, bone, connective tissue and central nervous system. IL-11 plays an important regulatory role in hematopoiesis by stimulating growth of myeloid, erythroid and megakaryocyte progenitor cells. It also regulates bone metabolism, inhibits production of proinflammatory cytokines, and protects against gastromucosal injury. Recombinant Human IL-11 is a 19.3 kDa protein consisting of 179 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.