Human Recombinant Nanog (from E. coli)
Supplier: Peprotech
120-21-5UG 120-21-20UG 120-21-100UG 120-21-250UG 120-21-500UG 120-21-1MG
10772-576EA 132.81 USD
10772-576 10772-578 10772-580 10772-582 10772-584 10772-586
Human Recombinant Nanog (from E. coli)
Proteins and Peptides
Nanog is a regulatory protein that is associated with undifferentiated pluripotent cells. The expression of nanog, which is suppressed in all adult tissues, is restricted to embryonic stem cells and to certain pluripotent cancer cells. Decreased expression of nanog is strongly correlated with cell differentiation. Nanog, most likely, acts as an intracellular regulator, that helps maintain pluripotency and self renewal via a STAT3-independent pathway. Recombinant Human Nanog is a 34.7 kDa protein, which is synthesized as a 304 amino acid polypeptide lacking a signal sequence for secretion.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.