Human Recombinant sCD22 (from CHO Cells)
Supplier: Peprotech
100-01-5UG 100-01-20UG 100-01-100UG 100-01-250UG 100-01-500UG 100-01-1MG
10770-618EA 132.81 USD
10770-618 10770-620 10770-622 10770-624 10770-626 10770-628
Human Recombinant sCD22 (from CHO Cells)
Proteins and Peptides
CD22 is a B-lineage restricted 135 kDa glycoprotein whose cell surface expression is limited to resting and activated B lymphocytes. The physiological role of CD22 is still unknown. Targeted disruption of CD22 in mice results in a reduced level of surface IgM on peripheral B cells, suggesting a role for CD22 in limiting antigen receptor signaling. CD22 is a member of the Ig gene superfamily that uniquely binds a sialic acid-dependent ligand. Recombinant Human sCD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.