Human Recombinant IL6 (from CHO Cells)
Supplier: ADIPOGEN CORP MS
CHI-HF-22006-C050 CHI-HF-22006-3050
102513-862EA 290.77 USD
102513-862 102513-864
Human Recombinant IL6 (from CHO Cells)
Proteins and Peptides
Interleukin 6 (IL-6) is a pleiotropic alpha-helical cytokine that plays important roles in acute phase reactions, inflammation, hematopoiesis, bone metabolism and cancer progression. IL-6 activity is essential for the transition from acute inflammation to either acquired immunity or chronic inflammatory disease. It is secreted by multiple cell types as a 22kDa-28kDa phosphorylated and variably glycosylated molecule. IL-6 induces signaling through a cell surface heterodimeric receptor complex composed of a ligand binding subunit (IL-6R) and a signal transducing subunit (gp130). IL-6 is a key factor for the growth of plasma cells.