Human Recombinant SuperFasLigand™ (Soluble)
Supplier: Enzo Life Sciences
522-020-3005 522-020-C005
89151-716EA 953.75 USD
89151-716 89151-718
Human Recombinant SuperFasLigand™ (Soluble)
Proteins and Peptides
Fas ligand.
Produced in HEK 293 cells. The extracellular domain of human FasL (APO-1L; CD95L; CD178) (aa 103-281) is fused at the N-terminus to a linker peptide (26 aa) and a FLAG®-tag. Glycosylation of rhsSuperFasLigandTM is similar to natural human FasL.