HDAC fluorometric deacetylase Substrate
Supplier: Enzo Life Sciences
260-137-M001 260-137-M005
89146-932EA 107.91 USD
89146-932 89146-934
HDAC fluorometric deacetylase Substrate
Proteins and Peptides
A fluorogenic substrate for histone deacetylase (HDAC). Ex(max): 330nm; Em(max): 395nm.