Ac-WEHD-CHO
Supplier: Enzo Life Sciences
260-055-M001 260-055-M005
89146-704EA 173.31 USD
89146-704 89146-706
Ac-WEHD-CHO
Proteins and Peptides
Inhibitor of caspase-1, -4 and -5
Reversible, cell permeable inhibitor of caspase-1 and -5. Also inhibits caspase-4.