Melittin
Supplier: Enzo Life Sciences
162-007-M001 162-007-MC05
89143-542EA 303.02 USD
89143-542 89143-544
Melittin
Proteins and Peptides
PLA2 activator
PLA2 stimulatory bee venom peptide. Sequence is similar to Phospholipase A2 Activating Peptide (Prod. No. ALX-165-007). Activates PLA2 (5-fold at 10µg/ml) in BC3H1 cells. For additional technical information see melittin (natural) (Prod. No. ALX-162-006).