Human Recombinant IL-1 alpha
Supplier: STEMCELL Technologies
78115 78115.1 78115.3
MSPP-78115EA 311.31 USD
MSPP-78115 MSPP-781151006 MSPP-781153
Human Recombinant IL-1 alpha
Proteins and Peptides
Interleukin 1 alpha (IL-1α) is a member of the IL-1 family and a dual-function cytokine. Both the unprocessed precursor and a processed IL-1α protein signal through IL-1 receptor type 1 (IL-1R1). Various cells, including keratinocytes, thymic epithelium, hepatocytes, endothelial cells, fibroblasts, and the epithelial cells of mucous membranes, have high levels of intracellular IL-1α precursor. The precursor is also expressed on the surface of monocytes and B lymphocytes (Netea et al.). IL-1α recruits infiltrating cells to a site of injury during necrosis and plays an important role during processes of sterile inflammation (Cohen et al.; Rider et al.). During hypoxia, IL-1α contributes to angiogenesis (Carmi et al.). Studies in mice show that IL-1α is produced by microglia-like cells after ischemic brain injury, which contributes to the inflammation (Luheshi et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').