Human Recombinant GRO-beta (CXCL2)
Supplier: STEMCELL Technologies
78112.1 78112.2
MSPP-781121006EA 738.08 USD
MSPP-781121006 MSPP-781122
Human Recombinant GRO-beta (CXCL2)
Proteins and Peptides
GRO (growth-regulated oncogene)-beta or CXCL2 is a member of the CXC family, which plays an integral role in recruitment and activation of neutrophils and basophils in response to tissue injury and microbial infection. GRO-beta and GRO-gamma are closely related to GRO-alpha and share 90% and 86% amino acid sequence homology, respectively, with GRO-alpha. Receptor-binding studies have demonstrated that GRO-alpha, -beta, and -gamma signal mainly through G protein-coupled receptors CXCR1 and CXCR2 (Ahuja and Murphy). GRO-beta is expressed in epithelial cells, monocytes, fibroblasts, and melanocytes and is further induced during inflammatory, epithelialization, and angiogenic processes, for example during the wound healing process of human burn wounds (Zaja-Milatovic and Richmond). GRO-beta also stimulates mitogenesis in certain human melanoma cells (Unemori et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').