SARS-CoV-2 (Spike Protein) Omicron XBB.1.5.X Peptide Pool
Supplier: STEMCELL Technologies
100-1422
MSPP-100-1422EA 1768.46 USD
MSPP-100-1422
SARS-CoV-2 (Spike Protein) Omicron XBB.1.5.X Peptide Pool
Proteins and Peptides
Stimulating T cells with SARS-CoV-2 (Spike Protein) Omicron XBB.1.5.X Mutation peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This pool is provided as two lyophilized mixtures (subpools) from the spike glycoprotein of SARS-CoV-2 Omicron XBB.1.5.X subvariant. The subpools contain 158 and 157 peptides, respectively, for a total of 315 peptides, and consist of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1269 on the spike protein. Through interactions with the angiotensin-converting enzyme 2 (ACE2) receptor, the spike protein enables the virus to attach to the cell membrane, and plays a critical role in viral entry (Hoffmann et al.; Walls et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').