Human Recombinant DcR3:Fc
Supplier: Enzo Life Sciences
522-055-C050
89151-790EA 597.32 USD
89151-790
Human Recombinant DcR3:Fc
Proteins and Peptides
Produced in HEK 293 cells. The extracellular domain of human DcR3 (decoy receptor 3) (aa 1-246) is fused to the Fc portion of human IgG1.