Leupeptin
Supplier: Enzo Life Sciences
260-009-M005 260-009-M025 260-009-M100
89146-576EA 56.68 USD
89146-576 89146-578 89146-580
Leupeptin
Proteins and Peptides
Cathepsin B inhibitor
Competitive and reversible inhibitor of serine and cysteine proteinases like calpain, porcine kallikrein, trypsin, plasmin, papain and cathepsin B. Does not inhibit chymotrypsin and thrombin. Also known to inhibit activation-induced programmed cell death and restoration of defective immune responses of HIV+ donors.
Order Now
SPECIFICATIONS
Protein/Peptide Name Leupeptin Leupeptin
Environmentally Preferable
Species
Source
Purity >97% (HPLC)