PROTEOSTAT® Protein Refolding and Aggregation Sensing Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
51040-KP002
10033-622EA 541.73 USD
10033-622
PROTEOSTAT® Protein Refolding and Aggregation Sensing Kit, Enzo Life Sciences
Assays Protein Assays
ProteoStat® Protein refolding and aggregation sensing kit provides a fractional factorial matrix design to facilitate screening of refolding parameters for a specific protein in different buffers, and a simple, homogenous assay format for monitoring protein aggregation using a red-emitting aggregation-sensitive dye. 