Accredited Thermometer Recertification & Calibration Services
Avantor Services offers recertification, calibration, and repair services for all makes and models of thermometers.
To order services for your instrument, please select the orange "Request a Quote Today" button.
Request a Quote Today Download Decontamination Form
Or, you can contact us at 1.888.793.2300 or services@avantorsciences.com.
Our thermometer services include:
- Complete visual inspection for any physical damage. If possible, the damage will be repaired.
- Recertification from 1 Point to 15 Points
- Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures
- Each temperature is recorded to 1/10th of a scale division
- A calibration report is generated, detailing pertinent information including the correction factors for each test point
- All calibration information is entered into a data storage system
- All certification procedures are performed in accordance with NIST and ASTM guidelines