VWR® Dry Block Heaters
VWR® Dry Block Heaters are compact, versatile, multi-purpose units available in a variety of block capacities with choices of digital and analog controls. All models offer exceptional temperature stability and uniformity. Digital models provide repeatable results and feature a temperature calibration mode. Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, and a variety of other laboratory procedures.
Each model accepts interchangeable modular blocks with over 40 options from which to choose. Blocks feature a close tube and block contact for maximum heat transfer and a thermometer well for measuring block temperature.
Product selection
VWR® Advanced Block Heaters
Our economical, versatile, and compact block heaters are designed for applications that require repeatable results and superior temperature stability.
VWR® Standard Block Heaters
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, and a variety of other laboratory procedures.
VWR® Advanced Dry Block Heater with Heated Lid
Provides exceptional temperature uniformity and stability, while reducing condensation on sample lids.
VWR® Block Heaters & Blocks
Modular blocks are constructed of anodized aluminum. The close contact of tube-block-walls allow for maximum heat retention.