High-quality labware for use in clinical laboratories
Introducing the Samco Clicktainer Vial
Thermo Scientific understands the value of patient samples. Sample collection needs to be easy. Sample transport needs to be secure. The Thermo Scientific™ Samco™ Clicktainer vials have a wide opening and leakproof seal. The audible “click” seal system indicates when the cap is properly applied so the sample is secure.
The Samco Clicktainer is ISO Compliant:
Tested in accordance with the requirements of BS EN 14254 and BS EN 14401 standards. View more versions of this product here.
*The term “leakproof” applies to Thermo Scientific Samco Clicktainer products, 90ml/53mm and 120ml/53mm, that meet the following criteria: a) a vacuum test that draws 95 kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.
Product selection
Samco Bio-Tite Specimen Containers
All plastic cap and vial transport system specifically designed for the collection and transport of fluids, solids and powders. The containers are ideal for the collection of urine, formalin or fixative fills and histology transport
Samco 24-hour Urine Collection Containers
Alleviate mess and exposure when performing urine collection and sampling. Amber tinted containers protect from light exposure and are inert to normal preservative
Samco Autoanalyzer Cups
Perfect choice for autosamplers, serum dilutions, microsampling and other laboratory applications. The conical wells are designed for complete sample pick up. Caps are available to minimize evaporation. Free standing base prevents tipping and spilling
Samco Disposable Culture Tubes
Designed for a wide range of laboratory applications. Each tube is precision molded and manufactured from premium grade materials. All sterile tubes are gamma-irradiated and ready to use
Samco Transfer Pipets
Expert craftsmanship produces the largest variety of transfer pipets in the industry. Designed to easily grip with latex gloves, the repeatable results will give you the confidence you need
Capitol Vial Specimen Collection
Sample collection is simplified with Capitol Vial Drugs of Abuse single and split collection kits. Complies with SAMHSA and DOT guidelines.