From Culture to Discovery
Ensure the most flexible, reproducible and reliable results across every stage of cell culture. From culture to cell modification, analysis and cryopreservation storage, Thermo ScientificTM Nalgene™ and Nunc™ cell culture products deliver consistent quality, performance and affordability – so researchers, like you, can advance their work with complete confidence.
Featured Product
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays.
Current Promotions
- Thermo Scientific™ Nunc™ 15 mL and 50 mL Conical Centrifuge Tubes - Buy 2 cases and receive one case of Serological Pipettes at no additional charge!
See all Thermo Scientific Promotions
Product selection
Cell Culture Dishes
Dishes in more than 100 combinations of format and surface. Excellent optical quality for manual or automated imaging
Cell Culture Flasks
Prepare and store culture media and perform a broad range of culturing techniques with our diverse selection of flat-sided flasks and accessories, plastic spinner flasks and plastic Erlenmeyer flasks
Cell Culture Microplates
Researchers all over the world choose Nunc microplates for their high quality, consistency and variety of available formats
Cell Culture Tubes
Choose from round and flat-bottom tubes as well as conical tubes that offer a variety of sizes and features, including recyclable racks and ease-of-use designs for enhanced lab performance
Conical Centrifuge Tubes
Take advantage of the improved separation and pelleting of a conical tube while eliminating risk of breakage with our polycarbonate and polystyrene tubes
Serological Pipettes
Advance your research with complete confidence. Featuring a full range of size and packaging options, our portfolio of serological pipettes offer the quality assurance and size variability you need
Specialty Coated
Make your most valuable work your best work with our special surface coatings. Our superior cell culture surfaces offer demonstrated performance that elevates stem cell and cell culture research