Which Preparation is Right for You?
Ward’s Preserved Specimens
The Widest Selection of Preparations, Backed by Expert Support
Compare all five of our preparation options to find the features that meet your needs. Click below each image for more details and to shop specimens.
Use the scroll bar below to scroll right for additional features.
Preparation Odor/Off-Gassing Formaldehyde/VOC Levels Fixation Injection Options Packaging Options Shelf Life Dissection Guides Support Additional Features/Benefits
Ward's Pure Preserved Specimens
High-quality fixation, low formalin odor
None
Low to no VOCs
(<1 ppm Acetone, Formaldehyde, Benzene, and other harmful chemicals)
Fully-Fixed
High quality tissue and clear differentiation of organs and structures
Plain
Single
Double
 Triple
Pails
Vacuum Packs
12-18 months unopened
After opening, several weeks or more
Free Dissection Guide Available Online
Backed by Ward's Science Plus Us team.
Ward's Science Exclusive Formalin-quality fixation without the odor or chemicals
No special ventilation or disposal process required
Formalin
Formalin Preserved Specimens
Highest-Quality for Hands-on Dissection
Some
As low as 1% formaldehyde - lowest possible levels to achieve maximum fixation results
Fully-fixed
Firm, sterile, thoroughly-preserved
Most true-to-life
Plain
Single
Double
Triple
Pails
Vacuum Packs
12-18 months unopened
After opening, several weeks or more
Free Dissection Guide Available Online
Backed by Ward's Science Plus Us team.
All specimens meet or exceed OSHA standards
Freeze-Dried
Freeze-Dried Specimens
Cost-effective and long-lasting
Some acetone odor from re-hydration
100% Chemical-Free
Organs and tissues retain natural coloration
Plain
Single
Double
Vacuum Packs
Unlimited shelf life in dries state
Re-hydrated specimens last for months
Free Dissection Guide Available Online
Backed by Ward's Science Plus Us team.
Ward's Science Exclusive
Lighter package weight for cost-effective shipping
Plastinated
Plastinated Specimens Permanently-Preserved for Easy Demonstration
None
100% Chemical-Free
Durable, dry, odorless, free of off-gassing
Real biological specimens that have been preserved, dissected, and permanently plastinated for demonstration and display.
Variety of injection options available
Varies
Unlimited; Built to last in your lab indefinitely.
N/A
Backed by Ward's Science Plus Us team.
Widest selection of platinated species available from any supplier